Assay Details
Target Gene Details
Entrez Gene ID: | 23845 |
Gene Name: | C-type lectin domain family 5, member a |
Gene Aliases: |
Clecsf5, Ly100, MDL-1, Mdl1 |
Location: |
Chr.6:40574898-40585805 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec5a | NM_001038604.1 | NP_001033693.1 | ||
| NM_021364.2 | NP_067339.1 | |||
| AF139769.1 | AAF02492.1 | |||
| AK036697.1 | BAC29537.1 | |||
| AK046600.1 | BAC32802.1 | |||
| AK137482.1 | BAE23374.1 | |||
| AK155409.1 | ||||
| BC104364.1 | AAI04365.1 | |||
| BC104365.1 | AAI04366.1 | |||
| BY205833.1 | ||||
| CN670295.1 |

Genomic Map