Assay Details
Target Gene Details
Entrez Gene ID: | 79362 |
Gene Name: | basic helix-loop-helix family, member e41 |
Gene Aliases: |
6430520M22Rik, Bhlhb2l, Bhlhb3, DEC2, Sharp1 |
Location: |
Chr.6:145858243-145865591 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bhlhe41 | NM_001271768.1 | NP_001258697.1 | ||
| NM_024469.2 | NP_077789.1 | |||
| AA013582.1 | ||||
| AB044090.1 | BAB21503.1 | |||
| AK137275.1 | 1 | 933 | ||
| AK170567.1 | 1 | 385 | ||
| BC037742.1 | AAH37742.1 | |||
| BE980915.1 | ||||
| BE985179.1 |
More Information
Set Membership: |
|

Genomic Map