Assay Details
Target Gene Details
Entrez Gene ID: | 208647 |
Gene Name: | cAMP responsive element binding protein 3-like 2 |
Gene Aliases: |
BBF2H7, C530025K05Rik, SCIRR69 |
Location: |
Chr.6:37327255-37442186 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Creb3l2 | NM_178661.4 | NP_848776.2 | ||
| AK035695.1 | BAC29155.1 | |||
| AK036712.1 | BAC29547.1 | |||
| AK041050.1 | BAC30798.1 | |||
| AK041669.1 | BAC31028.1 | |||
| AK042418.1 | ||||
| AK199399.1 | ||||
| BC043466.1 | AAH43466.1 | |||
| BC138470.1 | ||||
| BC138471.1 |

Genomic Map