Assay Details
Target Gene Details
Entrez Gene ID: | 28006 |
Gene Name: | family with sequence similarity 21 |
Gene Aliases: |
A130095H06, C530005J20Rik, D6Wsu116e |
Location: |
Chr.6:116208026-116262671 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 16 - Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam21 | NM_026585.3 | NP_080861.2 | ||
| AK038318.1 | BAC29966.1 | |||
| AK043764.1 | BAC31648.1 | |||
| AK052464.1 | 3 | 812 | ||
| AK122320.1 | BAC65602.2 | |||
| AK145716.1 | ||||
| AK161181.1 | ||||
| AK163765.1 | BAE37485.1 | |||
| AK168388.1 | ||||
| BC049979.1 | AAH49979.1 | |||
| BC056942.1 | AAH56942.1 |
More Information
Set Membership: |
|

Genomic Map