Assay Details
Target Gene Details
Entrez Gene ID: | 76223 |
Gene Name: | ATP/GTP binding protein-like 3 |
Gene Aliases: |
2900053G10Rik, 4930431N21Rik, 6530406M24Rik, CCP3 |
Location: |
Chr.6:34780432-34862184 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agbl3 | NM_001289656.1 | NP_001276585.1 | ||
| NM_001289657.1 | NP_001276586.1 | |||
| NM_178630.4 | NP_848745.2 | |||
| AK018326.1 | ||||
| AK029793.1 | BAC26620.1 | |||
| AK029798.1 | ||||
| AK030378.1 | BAC26932.1 | |||
| AK047649.1 | BAC33111.1 | |||
| AK054066.1 | ||||
| BC139065.1 | ||||
| DQ867031.1 | ||||
| DQ867032.1 |
More Information
Set Membership: |
|

Genomic Map