Assay Details
Target Gene Details
Entrez Gene ID: | 140494 |
Gene Name: | ATPase, H+ transporting, lysosomal V0 subunit A4 |
Gene Aliases: |
Atp6n1b, a4 |
Location: |
Chr.6:38048483-38124586 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v0a4 | NM_080467.3 | 16 | 2128 | NP_536715.3 |
| AB050903.1 | 15 | 1854 | BAB47243.1 | |
| AF326316.1 | 16 | 1922 | AAL30435.1 | |
| AF435090.1 | 16 | 2116 | AAN45855.1 | |
| AK128892.1 | BAC87655.1 | |||
| BC046979.1 | 17 | 2150 | AAH46979.1 |
More Information
Set Membership: |
|

Genomic Map