Assay Details
Target Gene Details
Entrez Gene ID: | 109624 |
Gene Name: | caldesmon 1 |
Gene Aliases: |
4833423D12Rik, AI195384, AV071549, AW536160, C920027I18Rik |
Location: |
Chr.6:34598500-34775469 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cald1 | NM_145575.3 | NP_663550.1 | ||
| AK050642.1 | ||||
| AK160593.1 | BAE35900.1 | |||
| BC019435.1 | AAH19435.1 | |||
| BC055908.1 | AAH55908.1 | |||
| BY308240.1 |
More Information
Set Membership: |
|

Genomic Map