Assay Details
Target Gene Details
Entrez Gene ID: | 12922 |
Gene Name: | corticotropin releasing hormone receptor 2 |
Gene Aliases: |
CRF-R2, CRFR2alpha, CRFR2beta, Crfr2 |
Location: |
Chr.6:55090048-55133016 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crhr2 | NM_001288618.1 | NP_001275547.1 | ||
| NM_001288619.1 | NP_001275548.1 | |||
| NM_001288620.1 | NP_001275549.1 | |||
| NM_009953.4 | NP_034083.2 | |||
| AK142261.1 | BAE24999.1 | |||
| AY445512.1 | AAS07021.1 | |||
| AY753668.1 | AAU94301.1 | |||
| BC137592.1 | ||||
| FJ548565.1 | ||||
| U17858.1 | AAA68026.1 | |||
| U19939.1 | AAC52243.1 | |||
| U21729.1 | AAC52174.1 |
More Information
Set Membership: |
|

Genomic Map