Assay Details
Target Gene Details
Entrez Gene ID: | 56449 |
Gene Name: | Y box protein 3 |
Gene Aliases: |
Csda, Dpba, MSY3, MSY4, Yb2, dbpA, oxyR |
Location: |
Chr.6:131364858-131388450 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ybx3 | NM_011733.2 | NP_035863.1 | ||
| NM_139117.2 | NP_620817.2 | |||
| AF246224.1 | AAF61741.1 | |||
| AF248546.1 | AAG14418.1 | |||
| AF248547.1 | AAG14419.1 | |||
| AK003145.1 | ||||
| AK076041.1 | BAC36141.1 | |||
| BC027785.1 | AAH27785.1 | |||
| BC048242.1 | AAH48242.1 | |||
| BC062377.1 | AAH62377.1 | |||
| D14485.1 | BAA03376.1 |

Genomic Map