Assay Details
Target Gene Details
Entrez Gene ID: | 78653 |
Gene Name: | bolA-like 3 (E. coli) |
Gene Aliases: |
1810056O20Rik |
Location: |
Chr.6:83349484-83358392 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bola3 | NM_175277.4 | NP_780486.1 | ||
| AK019037.1 | ||||
| AK028102.1 | BAC25747.1 | |||
| AK160488.1 | BAE35819.1 | |||
| AK218122.1 | ||||
| BC116947.1 | ||||
| BC116949.1 |

Genomic Map