Assay Details
Target Gene Details
Entrez Gene ID: | 12183 |
Gene Name: | 2,3-bisphosphoglycerate mutase |
Gene Aliases: |
AI323730, AL022789, C86192 |
Location: |
Chr.6:34476254-34505610 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bpgm | NM_007563.4 | NP_031589.1 | ||
| AK006315.1 | ||||
| AK037239.1 | ||||
| AK043412.1 | BAC31541.1 | |||
| AK078119.1 | BAC37133.1 | |||
| AK171685.1 | BAE42610.1 | |||
| AK211952.1 | ||||
| BC004589.1 | AAH04589.1 |
More Information
Set Membership: |
|

Genomic Map