Assay Details
Target Gene Details
Entrez Gene ID: | 109624 |
Gene Name: | caldesmon 1 |
Gene Aliases: |
4833423D12Rik, AI195384, AV071549, AW536160, C920027I18Rik |
Location: |
Chr.6:34598500-34775469 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cald1 | NM_145575.3 | 6 | 1274 | NP_663550.1 |
| AK148009.1 | 4 | 646 | BAE28286.1 | |
| BC019435.1 | 6 | 1233 | AAH19435.1 | |
| BQ769983.1 | 3 | 458 | ||
| CX568419.1 |
More Information
Set Membership: |
|

Genomic Map