Assay Details
Target Gene Details
Entrez Gene ID: | 17000 |
Gene Name: | lymphotoxin B receptor |
Gene Aliases: |
AI256028, CD18, LTbetaR, Ltar, TNF-R-III, TNFCR, TNFR-RP, TNFR2-RP, TNFRrp, Tnfbr, Tnfrsf3 |
Location: |
Chr.6:125306571-125313870 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ltbr | NM_010736.3 | NP_034866.1 | ||
| AK078859.1 | BAC37424.1 | |||
| AK146127.1 | BAE26920.1 | |||
| BC138590.1 | ||||
| BC138591.1 | ||||
| L38423.1 | AAB00846.1 | |||
| U29173.1 | AAA68964.1 |
More Information
Set Membership: |
|

Genomic Map