Assay Details
Target Gene Details
Entrez Gene ID: | 192196 |
Gene Name: | LUC7-like 2 (S. cerevisiae) |
Gene Aliases: |
4930471C18Rik, AA522013, AU015269, CGI59, Cgi74 |
Location: |
Chr.6:38551339-38609471 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Luc7l2 | NM_001170848.1 | NP_001164319.1 | ||
| NM_001170849.1 | NP_001164320.1 | |||
| NM_138680.2 | NP_619621.2 | |||
| AF318301.1 | AAK01182.1 | |||
| AK046307.1 | ||||
| AK134191.1 | BAE22044.1 | |||
| AK140271.1 | BAE24308.1 | |||
| AK147889.1 | BAE28206.1 | |||
| AK153527.1 | BAE32066.1 | |||
| BC020058.1 | ||||
| BC056354.1 | AAH56354.1 | |||
| BC056383.1 | AAH56383.1 | |||
| BC056970.1 | AAH56970.1 |

Genomic Map