Assay Details
Target Gene Details
Entrez Gene ID: | 14433 |
Gene Name: | glyceraldehyde-3-phosphate dehydrogenase |
Gene Aliases: |
Gapd |
Location: |
Chr.6:125161338-125166511 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
More Information
Set Membership: |
|

Genomic Map