Assay Details
Target Gene Details
Entrez Gene ID: | 18716 |
Gene Name: | prolactin induced protein |
Gene Aliases: |
GCDFP-15, GP17, mSMGP |
Location: |
Chr.6:41847549-41852062 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pip | NM_008843.3 | 4 | 492 | NP_032869.2 |
| AB017907.1 | 4 | 470 | BAA75745.1 | |
| AK138795.1 | 4 | 493 | BAE23780.1 | |
| BC028889.1 | 4 | 490 | AAH28889.1 | |
| S73282.1 | 4 | 488 | AAB31990.2 | |
| X02510.1 | 4 | 487 | CAA26346.1 |

Genomic Map