Assay Details
Target Gene Details
Entrez Gene ID: | 353172 |
Gene Name: | glycyl-tRNA synthetase |
Gene Aliases: |
GENA202, Gena201, Nmf249, Sgrp23 |
Location: |
Chr.6:55038001-55079504 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gars | NM_180678.3 | 16 | 2011 | NP_851009.2 |
| AK012758.2 | 16 | 2006 | ||
| AK146238.1 | 16 | 1988 | BAE27003.1 | |
| AK165857.1 | 16 | 1959 | BAE38417.1 | |
| AK182981.1 | 2 | 193 | ||
| AK190167.1 | 2 | 192 | ||
| AK191772.1 | 1 | 52 | ||
| AK198707.1 | 2 | 193 | ||
| AK198907.1 | 2 | 193 | ||
| AK200032.1 | 2 | 193 | ||
| BC021747.1 | 16 | 1992 | AAH21747.1 |
More Information
Set Membership: |
|

Genomic Map