Assay Details
Target Gene Details
Entrez Gene ID: | 353172 |
Gene Name: | glycyl-tRNA synthetase |
Gene Aliases: |
GENA202, Gena201, Nmf249, Sgrp23 |
Location: |
Chr.6:55038001-55079504 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gars | NM_180678.3 | NP_851009.2 | ||
| AK012758.2 | ||||
| AK146238.1 | BAE27003.1 | |||
| AK165857.1 | BAE38417.1 | |||
| AK183945.1 | ||||
| AK199850.1 | ||||
| AK211890.1 | ||||
| BC021747.1 | AAH21747.1 |
More Information
Set Membership: |
|

Genomic Map