Assay Details
Target Gene Details
Entrez Gene ID: | 21743 |
Gene Name: | indolethylamine N-methyltransferase |
Gene Aliases: |
Temt |
Location: |
Chr.6:55170627-55174990 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inmt | NM_009349.3 | 1 | 122 | NP_033375.1 |
| AK002281.1 | 1 | 170 | BAB21985.1 | |
| AK013010.1 | 1 | 159 | BAB28594.1 | |
| BC013518.1 | 1 | 148 | AAH13518.1 | |
| M88694.1 | 1 | 123 | AAA62365.1 |
More Information
Set Membership: |
|

Genomic Map