Assay Details
Target Gene Details
Entrez Gene ID: | 668210 |
Gene Name: | predicted gene 9047 |
Gene Aliases: |
EG668210 |
Location: |
Chr.6:29471437-29473429 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm9047 | NM_001145360.1 | 2 | 584 | NP_001138832.1 |
| BC147635.1 | 2 | 584 | ||
| BC147645.1 | 2 | 584 |
Target Gene Details
Entrez Gene ID: | 333088 |
Gene Name: | kielin/chordin-like protein |
Gene Aliases: |
AW060220, CRIM-2, Crim2, Gm793, KCP-1 |
Location: |
Chr.6:29473162-29508011 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcp | BC059909.1 | 12 | 2587 | AAH59909.1 |

Genomic Map