Assay Details
Target Gene Details
Entrez Gene ID: | 14695 |
Gene Name: | guanine nucleotide binding protein (G protein), beta 3 |
Gene Aliases: |
Gnb-3 |
Location: |
Chr.6:124834240-124840275 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnb3 | NM_013530.1 | NP_038558.1 | ||
| AK014789.1 | BAB29553.1 | |||
| AK030354.1 | 2 | 133 | BAC26917.1 | |
| BC018239.1 | AAH18239.1 |
More Information
Set Membership: |
|

Genomic Map