Assay Details
Target Gene Details
Entrez Gene ID: | 68794 |
Gene Name: | filamin C, gamma |
Gene Aliases: |
1110055E19Rik, ABP-280, ABPL, Fln2 |
Location: |
Chr.6:29433127-29461888 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Flnc | NM_001081185.1 | NP_001074654.1 | ||
| BC151097.1 | ||||
| BC158128.1 |

Genomic Map