Assay Details
Target Gene Details
Entrez Gene ID: | 232431 |
Gene Name: | G protein-coupled receptor, family C, group 5, member A |
Gene Aliases: |
Rai3, Raig1 |
Location: |
Chr.6:135065662-135084709 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gprc5a | NM_181444.5 | NP_852109.2 | ||
| BC036173.1 | AAH36173.1 | |||
| BC039217.1 | AAH39217.1 |
More Information
Set Membership: |
|

Genomic Map