Assay Details
Target Gene Details
Entrez Gene ID: | 231986 |
Gene Name: | JAZF zinc finger 1 |
Gene Aliases: |
AI591476, C820002C15, Jaz1, Tip27 |
Location: |
Chr.6:52768065-53068624 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jazf1 | NM_001168277.1 | NP_001161749.1 | ||
| NM_173406.3 | NP_775582.2 | |||
| AK043767.1 | BAC31649.1 | |||
| AK050474.1 | BAC34275.1 | |||
| BC048577.1 | AAH48577.1 |

Genomic Map