Assay Details
Target Gene Details
Entrez Gene ID: | 100042056 |
Gene Name: | RIKEN cDNA 9130019P16 gene |
Gene Aliases: |
6430501H15Rik, OTTMUSG00000023151 |
Location: |
Chr.6:54269681-54430221 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9130019P16Rik | NR_033635.1 | |||
| AK032212.1 | BAC27762.1 |

Genomic Map