Assay Details
Target Gene Details
Entrez Gene ID: | 27273 |
Gene Name: | pyruvate dehydrogenase kinase, isoenzyme 4 |
Gene Aliases: |
AV005916 |
Location: |
Chr.6:5483351-5496278 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdk4 | NM_013743.2 | NP_038771.1 | ||
| AJ001418.1 | CAA04752.1 | |||
| AK004543.1 | BAB23359.1 | |||
| AK036818.1 | BAC29590.1 | |||
| BC026134.1 | AAH26134.1 |
More Information
Set Membership: |
|

Genomic Map