Assay Details
Target Gene Details
Entrez Gene ID: | 317758 |
Gene Name: | GTPase, IMAP family member 9 |
Gene Aliases: |
A630002K24, BB145400 |
Location: |
Chr.6:48676121-48679116 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gimap9 | NM_174960.2 | NP_777620.1 | ||
| AK041336.1 | BAC30911.1 | |||
| BB618178.1 | ||||
| BC038043.1 | AAH38043.1 | |||
| BC096680.1 | AAH96680.1 |
More Information
Set Membership: |
|

Genomic Map