Assay Details
Target Gene Details
Entrez Gene ID: | 232087 |
Gene Name: | methionine adenosyltransferase II, alpha |
Gene Aliases: |
D630045P18Rik |
Location: |
Chr.6:72432796-72439608 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mat2a | NM_145569.4 | 1 | 112 | NP_663544.1 |
| AB070266.1 | 1 | 76 | BAD06937.1 | |
| AK034757.1 | 1 | 113 | BAC28823.1 | |
| AK050582.1 | 1 | 111 | ||
| AK052764.1 | 1 | 111 | BAC35139.1 | |
| AK079420.1 | 1 | 118 | BAC37642.1 | |
| AK133960.1 | 1 | 111 | BAE21955.1 | |
| AK144189.1 | 1 | 115 | BAE25756.1 | |
| AK145407.1 | 1 | 111 | BAE26420.1 | |
| AK150040.1 | 1 | 111 | BAE29262.1 | |
| AK151281.1 | 1 | 111 | BAE30267.1 | |
| AK160869.1 | 1 | 111 | ||
| AK161852.1 | 1 | 111 | BAE36605.1 | |
| AK164137.1 | 1 | 111 | BAE37643.1 | |
| AK166671.1 | 1 | 111 | BAE38932.1 | |
| AK168155.1 | 1 | 111 | BAE40120.1 | |
| AK169705.1 | 1 | 111 | BAE41317.1 | |
| BC003451.1 | 1 | 52 | AAH03451.1 | |
| BC058360.1 | 1 | 93 | AAH58360.1 |
Target Gene Details
Entrez Gene ID: | 78108 |
Gene Name: | promoter of Mat2a antisense radiation induced circulating long non-coding RNA |
Gene Aliases: |
4930414L22Rik, AI449819 |
Location: |
Chr.6:72438683-72440615 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Particl | NR_046011.1 | 1 | 765 | |
| AK145188.1 | 1 | 397 |
More Information
Set Membership: |
|

Genomic Map