Assay Details
Target Gene Details
Entrez Gene ID: | 232087 |
Gene Name: | methionine adenosyltransferase II, alpha |
Gene Aliases: |
D630045P18Rik |
Location: |
Chr.6:72432796-72439608 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mat2a | NM_145569.4 | NP_663544.1 | ||
| AB070266.1 | BAD06937.1 | |||
| AK034757.1 | BAC28823.1 | |||
| AK052764.1 | BAC35139.1 | |||
| AK079420.1 | BAC37642.1 | |||
| AK133960.1 | BAE21955.1 | |||
| AK144189.1 | BAE25756.1 | |||
| AK145407.1 | BAE26420.1 | |||
| AK150040.1 | BAE29262.1 | |||
| AK151281.1 | BAE30267.1 | |||
| AK160869.1 | ||||
| AK161852.1 | BAE36605.1 | |||
| AK164137.1 | BAE37643.1 | |||
| AK166671.1 | BAE38932.1 | |||
| AK168155.1 | BAE40120.1 | |||
| AK169705.1 | BAE41317.1 | |||
| AK190931.1 | ||||
| AK200511.1 | ||||
| AK206552.1 | ||||
| AK206774.1 | ||||
| AK210321.1 | ||||
| AK213948.1 | ||||
| AK215611.1 | ||||
| AK216312.1 | ||||
| AK219100.1 | ||||
| BC003451.1 | AAH03451.1 | |||
| BC058360.1 | AAH58360.1 |
More Information
Set Membership: |
|

Genomic Map