Assay Details
Target Gene Details
Entrez Gene ID: | 170772 |
Gene Name: | glucocorticoid induced transcript 1 |
Gene Aliases: |
2310047L21Rik, A130036A18Rik, Fam117c, Gig18, Tssn1 |
Location: |
Chr.6:8509600-8597549 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glcci1 | NM_001286728.1 | NP_001273657.1 | ||
| NM_001286729.1 | NP_001273658.1 | |||
| NM_133236.3 | NP_573499.2 | |||
| AK029524.1 | ||||
| AY098636.1 | AAM33072.1 | |||
| BC050802.1 | AAH50802.1 | |||
| CA561411.1 | ||||
| CN720421.1 |

Genomic Map