Assay Details
Target Gene Details
Entrez Gene ID: | 27056 |
Gene Name: | interferon regulatory factor 5 |
Gene Aliases: |
AW491843, mirf5 |
Location: |
Chr.6:29526130-29541851 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Irf5 | NM_001252382.1 | 3 | 544 | NP_001239311.1 |
| NM_012057.4 | 3 | 492 | NP_036187.1 | |
| AF028725.1 | 3 | 384 | AAB81997.1 | |
| AK150598.1 | 3 | 413 | ||
| AK151019.1 | 3 | 409 | BAE30037.1 | |
| AK152042.1 | 3 | 413 | ||
| AK156220.1 | 3 | 544 | BAE33631.1 | |
| AK171356.1 | 3 | 412 | ||
| BY749561.1 | 3 | 492 | ||
| BY749804.1 | 3 | 408 | ||
| EU401974.1 | 2 | 331 |
More Information
Set Membership: |
|

Genomic Map