Assay Details
Target Gene Details
Entrez Gene ID: | 16508 |
Gene Name: | potassium voltage-gated channel, Shal-related family, member 2 |
Gene Aliases: |
AI839615, AW555701, Kv4.2, R75121, mKIAA1044 |
Location: |
Chr.6:21216109-21729805 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnd2 | NM_019697.3 | 4 | 1614 | NP_062671.1 |
| AB093280.1 | 4 | 2095 | BAC41464.1 | |
| AF107780.1 | 4 | 1424 | AAD16972.1 | |
| AK032772.1 | 3 | 190 | BAC28015.1 | |
| BC079667.1 | 4 | 1614 | AAH79667.1 |
More Information
Set Membership: |
|

Genomic Map