Assay Details
Target Gene Details
Entrez Gene ID: | 243771 |
Gene Name: | poly (ADP-ribose) polymerase family, member 12 |
Gene Aliases: |
9930021O16, AA409132, AA536654, ARTD12, PARP-12, Zc3hdc1 |
Location: |
Chr.6:39086412-39118349 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp12 | NM_172893.3 | 11 | 1929 | NP_766481.2 |
| AK036886.1 | 11 | 1932 | BAC29622.1 | |
| AK053205.1 | 11 | 1830 | ||
| AK081528.1 | 11 | 1831 | ||
| AK156623.1 | 11 | 1798 | BAE33780.1 | |
| AK162200.1 | 11 | 1501 | ||
| AK178415.1 | 2 | 101 | ||
| BC024579.1 | 6 | 578 | AAH24579.1 | |
| BC028906.1 | 3 | 288 | AAH28906.1 | |
| BC048927.1 | 7 | 780 | AAH48927.1 | |
| BC120733.1 | 11 | 1762 | ||
| BC137645.1 | 11 | 1762 |

Genomic Map