Assay Details
Target Gene Details
Entrez Gene ID: | 243771 |
Gene Name: | poly (ADP-ribose) polymerase family, member 12 |
Gene Aliases: |
9930021O16, AA409132, AA536654, ARTD12, PARP-12, Zc3hdc1 |
Location: |
Chr.6:39086412-39118349 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp12 | NM_172893.3 | NP_766481.2 | ||
| AK036886.1 | BAC29622.1 | |||
| AK053205.1 | ||||
| AK081528.1 | ||||
| AK156623.1 | BAE33780.1 | |||
| AK162200.1 | ||||
| BC120733.1 | ||||
| BC137645.1 |

Genomic Map