Assay Details
Target Gene Details
Entrez Gene ID: | 18025 |
Gene Name: | nuclear factor, erythroid derived 2, like 3 |
Gene Aliases: |
Nrf3 |
Location: |
Chr.6:51432670-51458768 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfe2l3 | NM_010903.1 | NP_035033.1 | ||
| AB013852.1 | BAA76289.1 | |||
| AK017278.1 | 3 | 965 | BAB30669.1 | |
| AK133935.1 | BAE21936.1 | |||
| AK136980.1 | BAE23196.1 | |||
| AK137881.1 | BAE23511.1 | |||
| AK153612.1 | BAE32117.1 | |||
| BC005416.1 | AAH05416.1 |
More Information
Set Membership: |
|

Genomic Map