Assay Details
Target Gene Details
Entrez Gene ID: | 333088 |
Gene Name: | kielin/chordin-like protein |
Gene Aliases: |
AW060220, CRIM-2, Crim2, Gm793, KCP-1 |
Location: |
Chr.6:29473162-29508011 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 26 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcp | NM_001029985.4 | NP_001025156.3 | ||
| AK154369.1 | BAE32541.1 | |||
| AY884211.1 | AAX77677.1 |

Genomic Map