Assay Details
Target Gene Details
Entrez Gene ID: | 80782 |
Gene Name: | killer cell lectin-like receptor subfamily B member 1B |
Gene Aliases: |
Klrb1d, Ly55b, Ly55d, NKR-P1B, NKR-P1D, Nkrp1-b, Nkrp1d |
Location: |
Chr.6:128813706-128826375 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klrb1b | NM_030599.4 | NP_085102.4 | ||
| AF324826.1 | AAL37354.1 | |||
| AF338321.1 | AAK08512.1 | |||
| AF338322.1 | AAK08513.1 | |||
| AF342896.1 | AAK26639.1 | |||
| AK030702.1 | BAC27088.1 | |||
| AK041228.1 | BAC30869.2 | |||
| AK138766.1 | ||||
| AK169820.1 | BAE41390.1 | |||
| BC055379.1 | AAH55379.1 | |||
| BC100580.1 | AAI00581.1 | |||
| BC132126.1 | ||||
| BC132128.1 | ||||
| BC145389.1 | ||||
| DQ237928.1 | ||||
| M77677.1 | AAA39823.1 |
More Information
Set Membership: |
|

Genomic Map