Assay Details
Target Gene Details
Entrez Gene ID: | 17294 |
Gene Name: | mesoderm specific transcript |
Gene Aliases: |
AA408879, AI256745, Peg1 |
Location: |
Chr.6:30733506-30748466 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mest | NM_001252292.1 | NP_001239221.1 | ||
| NM_001252293.1 | NP_001239222.1 | |||
| NM_008590.2 | NP_032616.1 | |||
| AF482999.1 | AAM78507.1 | |||
| AK031718.1 | ||||
| AK032881.1 | BAC28068.1 | |||
| AK034949.1 | BAC28891.1 | |||
| AK146457.1 | ||||
| AK168714.1 | BAE40556.1 | |||
| AK168732.1 | BAE40573.1 | |||
| AK168743.1 | BAE40584.1 | |||
| AK168995.1 | BAE40794.1 | |||
| AK169095.1 | BAE40878.1 | |||
| AK169266.1 | BAE41027.1 | |||
| BC004019.1 | AAH04019.1 | |||
| BC006639.1 | AAH06639.1 | |||
| D16262.1 | BAA03795.1 |
More Information
Set Membership: |
|

Genomic Map