Assay Details
Target Gene Details
Entrez Gene ID: | 30853 |
Gene Name: | myeloid leukemia factor 2 |
Gene Aliases: |
AI256856, AU018944, C77284 |
Location: |
Chr.6:124931388-124936149 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mlf2 | NM_001170341.1 | NP_001163812.1 | ||
| NM_145385.2 | NP_663360.1 | |||
| AK141241.1 | BAE24610.1 | |||
| AK143972.1 | BAE25640.1 | |||
| BC003975.1 | AAH03975.1 |
More Information
Set Membership: |
|

Genomic Map