Assay Details
Target Gene Details
Entrez Gene ID: | 232408 |
Gene Name: | killer cell lectin-like receptor subfamily B member 1F |
Gene Aliases: |
A630024B12Rik, Nkr-p1f, Nkrp1-f, Nkrp1f |
Location: |
Chr.6:129036902-129059724 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klrb1f | NM_153094.2 | 1 | 68 | NP_694734.1 |
| NR_024262.1 | 1 | 68 | ||
| NR_024263.1 | 1 | 68 | ||
| AK041601.1 | 1 | 69 | BAC31001.1 | |
| AK154278.1 | 1 | 55 | BAE32484.1 | |
| AK170023.1 | 1 | 45 | BAE41516.1 | |
| AY029597.1 | AAK38158.1 | |||
| BC107177.1 | 1 | 49 | AAI07178.1 | |
| BC107178.1 | 1 | 49 | AAI07179.1 | |
| BC127141.1 | 1 | 49 | ||
| BC127142.1 | 1 | 45 | ||
| DQ237931.1 | ||||
| HQ713435.1 |
More Information
Set Membership: |
|

Genomic Map