Assay Details
Target Gene Details
Entrez Gene ID: | 107607 |
Gene Name: | nucleotide-binding oligomerization domain containing 1 |
Gene Aliases: |
C230079P11, Card4, F830007N14Rik, Nlrc1 |
Location: |
Chr.6:54923942-54972672 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nod1 | NM_172729.3 | NP_766317.1 | ||
| AK137585.1 | BAE23420.1 | |||
| BC042670.1 | AAH42670.1 |
More Information
Set Membership: |
|

Genomic Map