Assay Details
Target Gene Details
Entrez Gene ID: | 66881 |
Gene Name: | prenylcysteine oxidase 1 |
Gene Aliases: |
1200015P13Rik, AI115532, AI448340, PCL1, Pcly |
Location: |
Chr.6:86386006-86397158 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcyox1 | NM_025823.4 | 2 | 238 | NP_080099.1 |
| AK004799.2 | 2 | 193 | BAB23572.1 | |
| AK004840.1 | 2 | 193 | BAB23607.1 | |
| AK031585.1 | 2 | 194 | BAC27462.1 | |
| AK033373.1 | 2 | 194 | BAC28252.1 | |
| AK049412.1 | 2 | 191 | BAC33741.1 | |
| AK134528.1 | 2 | 239 | BAE22170.1 | |
| AK145366.1 | 2 | 198 | BAE26391.1 | |
| AK147190.1 | 2 | 206 | BAE27750.1 | |
| AK159981.1 | 2 | 194 | BAE35532.1 | |
| AK161616.1 | 2 | 194 | BAE36494.1 | |
| AK169209.1 | 2 | 194 | BAE40981.1 | |
| AK173058.1 | 2 | 160 | BAD32336.1 | |
| BC028308.1 | 2 | 208 | AAH28308.1 |
More Information
Set Membership: |
|

Genomic Map