Assay Details
Target Gene Details
Entrez Gene ID: | 66881 |
Gene Name: | prenylcysteine oxidase 1 |
Gene Aliases: |
1200015P13Rik, AI115532, AI448340, PCL1, Pcly |
Location: |
Chr.6:86386006-86397158 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcyox1 | NM_025823.4 | 3 | 468 | NP_080099.1 |
| AK004799.2 | 3 | 423 | BAB23572.1 | |
| AK004840.1 | 3 | 423 | BAB23607.1 | |
| AK031585.1 | 3 | 424 | BAC27462.1 | |
| AK033373.1 | 3 | 424 | BAC28252.1 | |
| AK049412.1 | 3 | 421 | BAC33741.1 | |
| AK134528.1 | 3 | 469 | BAE22170.1 | |
| AK145366.1 | 3 | 428 | BAE26391.1 | |
| AK147190.1 | 3 | 436 | BAE27750.1 | |
| AK159981.1 | 3 | 424 | BAE35532.1 | |
| AK161616.1 | 3 | 424 | BAE36494.1 | |
| AK169209.1 | 3 | 424 | BAE40981.1 | |
| AK173058.1 | 3 | 390 | BAD32336.1 | |
| BC006056.1 | 1 | 79 | AAH06056.1 | |
| BC028308.1 | 3 | 438 | AAH28308.1 |
More Information
Set Membership: |
|

Genomic Map