Assay Details
Target Gene Details
Entrez Gene ID: | 15258 |
Gene Name: | homeodomain interacting protein kinase 2 |
Gene Aliases: |
1110014O20Rik, B230339E18Rik, Stank |
Location: |
Chr.6:38687512-38876190 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hipk2 | NM_001136065.2 | 14 | 3219 | NP_001129537.1 |
| NM_001294143.1 | 14 | 3298 | NP_001281072.1 | |
| NM_001294144.1 | 14 | 3295 | NP_001281073.1 | |
| NM_010433.2 | 14 | 3379 | NP_034563.2 | |
| AF077659.1 | 14 | 3142 | AAC63011.1 | |
| AF170301.1 | 14 | 3048 | AAD52566.1 | |
| AF170302.1 | 14 | 3129 | AAD52567.1 | |
| AF208292.1 | 14 | 3132 | AAG41237.1 | |
| AF273680.1 | 14 | 3019 | AAG02078.1 | |
| AF333791.1 | 14 | 2922 | AAK07649.1 | |
| AF333792.1 | 14 | 3006 | AAK07650.1 | |
| BC031904.1 | 14 | 3031 | AAH31904.1 |

Genomic Map