Assay Details
Target Gene Details
Entrez Gene ID: | 13619 |
Gene Name: | polyhomeotic-like 1 (Drosophila) |
Gene Aliases: |
AW557034, Edr, Edr1, Mph1, Rae-28, rae28 |
Location: |
Chr.6:122317731-122340227 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phc1 | NM_001042623.2 | NP_001036088.1 | ||
| NM_001271579.1 | NP_001258508.1 | |||
| NM_007905.3 | NP_031931.2 | |||
| NR_073364.1 | ||||
| AK132756.1 | BAE21337.1 | |||
| AK167590.1 | BAE39648.1 | |||
| BC046535.1 | AAH46535.1 | |||
| BC052394.1 | AAH52394.1 | |||
| S73882.1 | AAB31766.1 | |||
| S73883.1 | ||||
| S73884.1 | ||||
| U63386.1 | AAC28974.1 |
More Information
Set Membership: |
|

Genomic Map