Assay Details
Target Gene Details
Entrez Gene ID: | 243371 |
Gene Name: | leucine rich repeat containing 61 |
Gene Aliases: |
BC027309 |
Location: |
Chr.6:48554779-48570722 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc61 | NM_001110160.1 | NP_001103630.1 | ||
| NM_177736.3 | 1 | 81 | NP_808404.2 | |
| AK136901.1 | 1 | 67 | BAE23163.1 | |
| AK143222.1 | 1 | 84 | BAE25315.1 | |
| AK170792.1 | ||||
| AK171463.1 | BAE42469.1 | |||
| BY023081.1 | ||||
| CV563372.1 |

Genomic Map