Assay Details
Target Gene Details
Entrez Gene ID: | 194357 |
Gene Name: | monooxygenase, DBH-like 2 |
Gene Aliases: |
Dbhl1 |
Location: |
Chr.6:40878794-40887494 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Moxd2 | NM_139296.2 | NP_647457.2 | ||
| AB065134.1 | BAB62024.1 |
More Information
Set Membership: |
|

Genomic Map