Assay Details
Target Gene Details
Entrez Gene ID: | 56524 |
Gene Name: | membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6) |
Gene Aliases: |
P55t, Pals2 |
Location: |
Chr.6:50110241-50198598 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpp6 | NM_001164733.1 | 1 | 141 | NP_001158205.1 |
| NM_001164734.1 | 1 | 141 | NP_001158206.1 | |
| NM_019939.2 | 1 | 141 | NP_064323.2 | |
| AF161181.1 | AAD45009.1 | |||
| AF199009.1 | 1 | 137 | AAF63790.1 | |
| AF199010.1 | 1 | 137 | AAF63791.1 | |
| AK144434.1 | 1 | 142 | BAE25887.1 | |
| BC046421.1 | 1 | 88 | AAH46421.1 | |
| BC069908.1 | 1 | 130 | AAH69908.1 | |
| BC106140.1 | 1 | 88 | AAI06141.1 | |
| BC138666.1 | ||||
| BC145363.1 | ||||
| BC145364.1 |
More Information
Set Membership: |
|

Genomic Map