Assay Details
Target Gene Details
Entrez Gene ID: | 56524 |
Gene Name: | membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6) |
Gene Aliases: |
P55t, Pals2 |
Location: |
Chr.6:50110241-50198598 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpp6 | NM_001164733.1 | 12 | 1539 | NP_001158205.1 |
| NM_001164734.1 | 12 | 1549 | NP_001158206.1 | |
| NM_019939.2 | 11 | 1507 | NP_064323.2 | |
| AF161181.1 | 11 | 1371 | AAD45009.1 | |
| AF199009.1 | 11 | 1503 | AAF63790.1 | |
| AF199010.1 | 12 | 1545 | AAF63791.1 | |
| AK077754.1 | 2 | 125 | BAC36993.1 | |
| AK144434.1 | 12 | 1550 | BAE25887.1 | |
| AK184594.1 | 2 | 200 | ||
| AK187690.1 | 3 | 347 | ||
| BC138666.1 | 11 | 1365 | ||
| BC145363.1 | 12 | 1407 | ||
| BC145364.1 | 12 | 1397 |
More Information
Set Membership: |
|

Genomic Map