Assay Details
Target Gene Details
Entrez Gene ID: | 109648 |
Gene Name: | neuropeptide Y |
Gene Aliases: |
0710005A05Rik |
Location: |
Chr.6:49822729-49829507 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npy | NM_023456.2 | NP_075945.1 | ||
| AF273768.1 | AAG00945.1 | |||
| AF286198.1 | AAG01330.1 | |||
| AF286199.1 | AAG01331.1 | |||
| AK002982.1 | BAB22495.1 | |||
| BC043012.1 | AAH43012.1 |
More Information
Set Membership: |
|

Genomic Map